Learn More
Medchemexpress LLC Apronal | 528-92-7 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB217710MM/1ML
Encompass_Preferred
Apronal (Allylisopropylacetylurea, Apronalide) is utilized in the research of neuropsychiatry disorders. Research indicates that allylisopropylacetylurea elevates the levels of porphyrins in the liver and urine of animals, also causing a distinct green pigmentation of the liver. Animals treated with high doses over long periods exhibit good recovery, with measured parameters returning to normal values.
  • Used for neuropsychiatry research
  • Elevates porphyrins in liver and urine
  • Causes green pigmentation of the liver
  • Long-term treatment is reversible with good recovery
Catalog No. 50-004-12405