Learn More
Ubpbio A 58 amino acid small protein that is a potent serine protease inhibitor. CAS# 9087-70-1, ≥3.0 EPU/mg
Supplier:  Ubpbio P105320
Encompass_Preferred
Aprotinin is a 58 amino acid small protein derived from bovine lung tissue that is highly positively charged and contains three disulfide bonds. It is a competitive inhibitor of several serine proteases, including trypsin, chymotrypsin, thrombin, and plasmin. In clinical, it is used as an anti-clotting agent for reducing bleeding. This product is for research use only. Typical working concentration is 2 µg/ml. 
Catalog No. 50-233-9288