Learn More
TARGETMOL CHEMICALS INC APROTININ 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T335925MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Aprotinin (Traskolan) a broad-spectrum serine protease inhibitor inhibiting the activity of a number of different esterases and proteases including trypsin chymotrypsin kallikrein plasmin tissue plasminogen activator and tissue and leukocytic proteinases. purity: 98%
Catalog No. 50-257-6959