Learn More
Research Products International Corp Aprotinin, Bovine Lung, 10 Milligrams
SDP
Supplier:  Research Products International Corp A205500.01
Encompass_Preferred
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. The binding is reversible, and most aprotinin-protease complexes dissociate at a pH>10 or <3.2. Target enzumes include trypsin, chymatrypsin, kalikrein and plasmin.
Activity: >4500 KIU/mg
Catalog No. 50-550-388