Learn More
Medchemexpress LLC Aprutumab 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP990075MG
Encompass_Preferred
Aprutumab (BAY 1179470) is a fully human FGFR2 monoclonal antibody which binds to the FGFR2 isoforms FGFR2-IIIb and FGFR2-IIIc Aprutumab has the potential for solid tumors research[1]
Catalog No. 50-003-24886