Learn More
Medchemexpress LLC HY-124944 5mg Medchemexpress, APS6-45 CAS:2188236-41-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1249445MG
Encompass_Preferred
Medchemexpress, HY-124944 5mg APS6-45 CAS:2188236-41-9 APS6-45 is an orally active tumor-calibrated inhibitor (TCI). APS6-45 inhibits RAS/MAPK signaling and exhibits antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1059