Learn More
Medchemexpress LLC Aptiganel hydrochloride | 137160-11-3 | 99.9% | 339.86 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY110097100MG
Encompass_Preferred
Aptiganel hydrochloride is a non-competitive NMDA receptor antagonist with neuroprotective effects. It is supplied as a solid, off-white to light yellow compound primarily for research purposes.
  • Targets iGluR and NMDA receptor
  • Involved in membrane transporter/ion channel and neuronal signaling pathways
  • Powder stable for 3 years at -20°C or 2 years at 4°C
  • Solution stable for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-003-43487