Learn More
Selleck Chemical LLC Apv-527-A4057-1MG
SDP
Supplier:  Selleck Chemical LLC A40571MG
Encompass_Preferred
APV-527 is a bispecific antibody targeting 4-1BB on T-cells and the 5T4 tumor antigen to stimulate immune responses against solid tumors It is being developed for advanced solid tumors including non-small cell lung cancer breast cancer and pancreatic cancer with positive safety and efficay M W 157 94 kDa
Catalog No. 50-313-8472