Learn More
Medchemexpress LLC HY-120801A 5mg Medchemexpress, APX-115 (free base) CAS:1270084-92-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120801A5MG
Encompass_Preferred
Medchemexpress, HY-120801A 5mg APX-115 (free base) CAS:1270084-92-8 APX-115 free base (Ewha-18278 free base) is a potent, orally active pan NADPH oxidase (Nox) inhibitor with Ki values of 1.08 μM, 0.57 μM, and 0.63 μM for Nox1, Nox2 and Nox4, respectively. APX-115 free base effectively prevents kidney injury. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9613