Learn More
Selleck Chemical LLC APX2009 E1987-5MG
SDP
Supplier:  Selleck Chemical LLC E19875MG
Encompass_Preferred
APX2009 is a second-generation redox-specific inhibitor of APE1/Ref-1 an essential enzyme in the base excision repair (BER) pathway APX2009 effectively reduces breast cancer cell proliferation and colony formation
Catalog No. 50-000-39241