Learn More
Sigma Aldrich Fine Chemicals Biosciences Apyrase from potato recomb
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A6237500UN
Encompass
Apyrase also known as ATP-diphosphohydrolase is expressed in wide variety of plant and animal tissues. The commercially accessible apyrase enzyme is isolated from potato tubers.
Catalog No. 50-290-5323