Learn More
Cayman Chemical aPyrrolidinopropiophenone hy
SDP
Supplier:  Cayman Chemical 104455 MG
Encompass_Preferred
An analog of the appetite suppressant diethylcathinone related to the designer drug 4'-methyl-α-pyrrolidinopropiophenone (MPPP); metabolized to a variety of products, including cathinone; intended to be used for forensic applications
Catalog No. 50-289-1330