Learn More
Electron Microscopy Sciences Aqueous Glutaraldehyde 50% in Serum, EM Grade 10 ML
SDP
Supplier:  Electron Microscopy Sciences 16316
Encompass_Preferred
Over the years, many of our valued customers have requested purchasing our aqueous Glutaraldehyde in serum vials. This packaging allows the user to puncture the special non-reactive seal with a hypodermic syringe and draw out the exact quantity of aqueous Glutaraldehyde needed without opening the container. This packaging assures freshness and purity of the aqueous Glutaraldehyde at all times.
Grade: EM Grade
Catalog No. 50-285-96