Learn More
AdipoGen AQC, 148757-94-2, MFCD01940654, 250 mg
SDP
Supplier:  AdipoGen CDXA0057M250
Encompass_Preferred
MF: C14H11N3O4, MW: 285.25, ≥95%, Appearance: White to off-white crystalline powder. Suitable for amino acid or protein sequence analysis by HPLC with fluorescence detection.
Catalog No. 50-595-786