Learn More
Foxx Life Sciences AQC 3/8 HB NS AQC22006HT 5PK
Supplier:  Foxx Life Sciences AQC22006HT5
Encompass_Preferred
Manufactured by CPC AseptiQuik C Connectors provide quick and easy sterile connections even in non-sterile environments AseptiQuik C connectors CLICK-PULL-TWIST design enables users to transfer media easily with less risk of operator error The connectors robust design provides reliable performance without the need for clamps fixtures or tube welders Biopharmaceutical manufacturers can make sterile connections with the quality and market availability they expect from the leader in single-use connection Features CLICK-PULL-TWIST Design Intuitive three-step connection process that reduces the risk of operator error Enable simultaneous and secure removal of both membranes for a reliable sterile connection Delivers repeatable dependable performance without the need for additional hardware Integrated Lock Ring Secures the final connection and prevents accidental disassembly CPC Click Confirmation Provides an audible click to confirm proper and complete assembly
Catalog No. 50-005-72971