Learn More
Medchemexpress LLC HY-117695 50mg Medchemexpress, AQC CAS:148757-94-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11769550MG
Encompass_Preferred
Medchemexpress, HY-117695 50mg AQC CAS:148757-94-2 AQC (6-Aminoquinolyl-N-hydroxysccinimidyl carbamate) is a reagent used for amino acid or protein sequence analysis by HPLC with fluorescence detection. AQC reacts with primary and secondary amino acids to yield fluorescent derivates, allowing amino acid detection at under-picomolar levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0907