Learn More
Waters Corp Waters Alliance Standards, Qualification, 10 mL, Fluorescence
Supplier: Waters Corp 700002753
This HPLC standards kit, for system assessment, includes 8 ampules of 10 mL standard solutions with varying Anthracene quantities and a blank diluent. Prepared in 20% 18 M ASTM Type 1 Lab Water (± 1%) and 80% HPLC Grade Acetonitrile (± 1%) V/V, it includes a Certificate of Analysis, ensuring high injection precision and reliable retention time.
- Designed for system assessment and qualification.
- Kit includes 8 ampules, 10 mL each.
- Contains Anthracene standards and a blank diluent.
- Certificate of Analysis provided.
- Ensures high injection precision and accurate quantification.
- Provides exceptional retention time.
- Uses Reversed Phase separation and Fluorescence detection.
- Liquid formulation, stored at 5°C ± 3°C (Fridge).
- Shelf life: 24 months.
- Hazardous material, special freight handling required.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.