Learn More
Medchemexpress LLC AQX-016A 25mg | 849669-54-1 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11562025MG
Encompass_Preferred
AQX-016A is an orally active SHIP1 agonist research compound intended for laboratory use. It activates recombinant SHIP1 in vitro, modulates PI3K signaling, and is used as a chemical tool for inflammation and cell signaling studies. Provided as a solid with defined solubility and storage recommendations.
  • High purity (≈99.5%) suitable for biochemical assays.
  • Small-molecule SHIP1 agonist for modulation of PI3K pathway.
  • Soluble in DMSO and ethanol with ultrasonic/warming recommended.
  • Stable as powder at -20°C for long-term storage.
  • Packaged as 25 MG for laboratory testing.
Catalog No. 50-002-28911