Learn More
Medchemexpress LLC HY-13248 1mg Medchemexpress, AR-C155858 CAS:496791-37-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132481MG
Encompass_Preferred
Medchemexpress, HY-13248 1mg AR-C155858 CAS:496791-37-8 AR-C155858 is a selective monocarboxylate transporter MCT1 and MCT2 inhibitor with Kis of 2.3 nM and 10 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8969
 Restricted Item