Learn More
Medchemexpress LLC HY-101039A 5mg Medchemexpress, AR-M 1000390 (hydrochloride) CAS:209808-47-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101039A5MG
Encompass_Preferred
Medchemexpress, HY-101039A 5mg AR-M 1000390 (hydrochloride) CAS:209808-47-9 AR-M 1000390 hydrochloride is an exceptionally selective, potent δ opioid receptor agonist with an EC 50 of 7.2±0.9 nM for δ agonist potency. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2018