Learn More
Medchemexpress LLC AR-M 1000390 (hydrochloride) | 209808-47-9 | 98.2% | 384.94 g/mol | C23H29ClN2O | 1 ML

Supplier: Medchemexpress LLC HY101039A10MM 1ML
AR-M 1000390 hydrochloride is a selective, potent δ-opioid receptor agonist used as a research tool in opioid receptor pharmacology. It exhibits sub-10 nM in vitro potency and is supplied as a concentrated solution suitable for receptor assays and signaling studies.
- Selective δ-opioid receptor agonist with EC50 7.2 ± 0.9 nM.
- Supplied as a 10 mM solution in DMSO, 1 mL vial.
- High purity (98.2%) suitable for biochemical assays.
- CAS number 209808-47-9 for unambiguous identification.
- Soluble in DMSO and water; in vivo vehicle protocols available.
- Molecular weight of the hydrochloride salt: 384.94 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.