Learn More
Medchemexpress LLC AR420626 10mM 1mL | 1798310-55-0 | 417.29 | C21H18Cl2N2O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11652210MM 1ML
Encompass_Preferred
AR420626 is a selective agonist of free fatty acid receptor 3 (FFAR3) supplied for research use. It is offered as a 10 mM solution in DMSO (1 mL) and is reported to have high purity, making it suitable for in vitro pharmacology and signaling pathway studies.
  • Selective FFAR3 agonist with reported nanomolar activity
  • Supplied as a 10 mM solution in DMSO (1 mL)
  • High reported purity (approximately 98.3%)
  • Suitable for in vitro pharmacology and signaling studies
  • Available as solid for custom preparations
Catalog No. 50-002-29261