Learn More
Percival Scientific Arabidopsis Growth Chamber, AR36L3H15, 29.7 cu. ft., SciWhite with red LEDs, 3 tiers, 7-44° C ±0.5°, IntellusUltra controller, touchscreen, ultrasonic humidifier and dehumidifier
SDP
Supplier:  Percival Scientific AR36L3H15
Encompass_Preferred
The AR36L3H15 growth chamber is designed for growth of Arabidopsis and low light plant growth applications. SciWhite with red LEDs are dimmable from 10-100% output with up to 350 µmoles/m2/s irradiance @ 6". Ultrasonic humidifier, dehumidifier, standard humidity sensor provide humidity control. Constructed of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Special airflow insulates shelf level experiments from heat generated by underlying light fixtures. Three epoxy coated steel wire shelves slide easily on stainless steel rail assemblies. Self-contained air-cooled condensing unit with continuous compressor operation extends life, ensuring precise temperature control by cycling refrigerant and hot gas to coil; prolonging life of compressor, eliminating risk of ice build up in coil. Top mounted refrigeration system allows easy access for service. Contact sales@percival-scientific.com for more options.
Catalog No. NC3794489
Only null left
Add to Cart
Request a Quote