Learn More
Medchemexpress LLC Arabinogalactan from larch wood | 9036-66-2 | MFCD00062638 | 99.6% | 500.49144 g/mol | C20H36O14 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW350004100G
Encompass_Preferred
Arabinogalactan from larch wood is a natural polysaccharide reagent commonly used in glycobiology and carbohydrate-related biochemical research. Supplied as an off-white to light-yellow powder with a high assay, it is suitable for use as a reagent component in assays and formulation work. Recommended storage is powder at -20°C; in solvent, store at -80°C for up to 6 months or at -20°C for up to 1 month.
  • Natural polysaccharide extracted from larch wood.
  • High assay purity (≈99.6%).
  • Off-white to light-yellow powder, easy to weigh and handle.
  • Suitable for glycobiology and carbohydrate-related biochemical assays.
  • Stable when stored as powder at -20°C; in solution store at -80°C or -20°C per recommended durations.
  • Available in laboratory pack sizes, including 100 g.
Catalog No. 50-002-82120