Learn More
Medchemexpress LLC Arabinose-d | 77583-92-7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN2353S1100MG
Encompass_Preferred
Arabinose-d is the deuterium labeled Arabinose. Arabinose is an endogenous metabolite. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation
Catalog No. 50-004-20374