Learn More
Medchemexpress LLC Arachidic acid-d39 | 39756-32-6 | 351.77 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW004260S1
Encompass_Preferred
Arachidic acid-d39 is the deuterium labeled Arachidic acid. Arachidic acid (Eicosanoic acid) is a long-chain saturated fatty acid found in human plasma phospholipid, fish, cannabis, and other plant oils. It can be used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled, which can be used as a tracer for quantitation during the drug development process.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Suitable for anti-cancer drug delivery research when conjugated with Chitosan oligosaccharide.
  • Available for research use only.
Catalog No. 50-004-46697