Learn More
Medchemexpress LLC Arachidic acid-d4 | 1219803-69-6 | 316.56 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW004260S45MG
Encompass_Preferred
Arachidic acid-d4 is a deuterium-labeled version of Arachidic acid (Eicosanoic acid), a long-chain saturated fatty acid found in human plasma phospholipid, fish, cannabis, and other plant oils.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
  • Conjugated with Chitosan oligosaccharide, it can be used in anti-cancer drug delivery research
Catalog No. 50-004-45614