Learn More
Medchemexpress LLC Arachidic acid-d4 (Icosanoic acid-d4) | 1219803-69-6 | 316.56 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW004260S410MG
Encompass_Preferred
Arachidic acid-d4 is the deuterium labeled Arachidic acid (Eicosanoic acid), a long-chain saturated fatty acid found in human plasma phospholipid, fish, cannabis, and other plant oils. Deuterium labeling allows for its use as a tracer and internal standard in quantitative analysis. Deuteration affects the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporated into drug molecules as tracers for quantitation during drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-45851