Learn More
Medchemexpress LLC Arachidic acid (Eicosanoic acid) | 506-30-9 | 312.53 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0042601G
Encompass_Preferred
Arachidic acid is an orally bioavailable membrane fluidity modulator and receptor/ion channel effector. It does not activate arachidonic acid-sensitive K+ channels in rat midbrain and hypothalamic neurons. Arachidic acid has been shown to alleviate age-related spatial cognitive impairment and is applicable for research related to this condition.
  • Orally bioavailable
  • Membrane fluidity modulator
  • Receptor/ion channel effector
  • Does not activate arachidonic acid-sensitive K+ channels
  • Alleviates age-related spatial cognitive impairment
Catalog No. 50-004-47470