Learn More
Cayman Chemical ArachIdonIc ACD13C5 mthyl 50ug
SDP
Supplier:  Cayman Chemical 1746150 UG
Encompass_Preferred
An internal standard for the quantification of arachidonic acid methyl ester by GC- or LC-MS
Catalog No. 50-296-5502