Learn More
Cayman Chemical ArachIdonIc ACDd11 ethyl S
SDP
Supplier:  Cayman Chemical 37970100 UG
Encompass_Preferred
An internal standard for the quantification of arachidonic acid ethyl ester by GC- or LC-MS
Catalog No. 50-291-6358