Learn More
Cayman Chemical Arachidonic Acid Alkyne
SDP
Supplier:  Cayman Chemical 10538100 UG
Encompass_Preferred
A form of arachidonic acid with an ω-terminal alkyne; can be used in click chemistry linking reactions to tag arachidonic acid as well as metabolites and derivatives
Catalog No. 50-288-8094