Learn More
Cayman Chemical ArachIdonIc AcId-d5 100UG
SDP
Supplier:  Cayman Chemical 9000477100 UG
Encompass_Preferred
An internal standard for the quantification of arachidonic acid by GC- or LC-MS
Catalog No. 50-289-2442