Learn More
Cayman Chemical ArachIdonIc AcId-d8 5mg
SDP
Supplier:  Cayman Chemical 3900105 MG
Encompass_Preferred
An internal standard for the quantification of arachidonic acid by GC- or LC-MS
Catalog No. 50-291-5714