Learn More
Cayman Chemical Arachidonic Acid ethyl ester
SDP
Supplier:  Cayman Chemical 1000820050 MG
Encompass_Preferred
An esterified form of arachidonic acid; commonly used as a source of exogenous arachidonic acid in vivo; has been found in the meconium of newborns suspected of fetal alcohol exposure
Catalog No. 50-288-8681