Learn More
Medchemexpress LLC Arachidonic acid sodium salt | 6610-25-9 | 99.0% | 326.45 g/mol | C20H31NaO2 | 5 MG

Supplier: Medchemexpress LLC HY109590A5MG
Arachidonic acid sodium salt is the monosodium salt of the polyunsaturated omega-6 fatty acid arachidonic acid, supplied as a research-grade solid for use as a substrate in lipid signaling, eicosanoid biosynthesis assays, and analytical workflows. Handle under inert atmosphere and protect from light for optimal stability.
- Monosodium salt form of arachidonic acid.
- CAS 6610-25-9.
- Molecular weight 326.45 g/mol.
- Purity ~99.0% by supplier specification.
- Offered in small research sizes including 5 mg.
- Store at -20°C, protect from light, under nitrogen; in solution store at -80°C for long term.
- Suitable as substrate for lipid signaling and metabolic enzyme assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.