Learn More
Cayman Chemical ArachidonoylN Ndimethyl amide
SDP
Supplier:  Cayman Chemical 100072935 MG
Encompass_Preferred
An analog of AEA that exhibits weak or no binding to the human CB1 receptor (Ki >1 µM); inhibits rat glial gap junction cell-cell communication at a concentration of 50 µM
Catalog No. 50-289-0944