Learn More
Cayman Chemical ArachIdonyl 2CLOroethylam 10mg
SDP
Supplier:  Cayman Chemical 9105410 MG
Encompass_Preferred
A potent and selective CB1 receptor agonist (Kis = 1.4 and >2,000 nM for CB1 and CB2 receptors, respectively); induces hypothermia in mice with the same efficacy as AEA in spite of its higher affinity for the CB1 receptor and thus is a possible substrate for FAAH
Catalog No. 50-289-4868