Learn More
Cayman Chemical ArachIdonyl Alcohol 10mg
SDP
Supplier:  Cayman Chemical 3253910 MG
Encompass_Preferred
A polyunsaturated fatty alcohol; been used as a substrate in the synthesis of various ether lipids
Catalog No. 50-294-6786