Learn More
Cayman Chemical ArachIdonyl-L-carnItInchl 10mg
SDP
Supplier:  Cayman Chemical 900093910 MG
Encompass_Preferred
An acylcarnitine formed from carnitine conjugated to arachidonic acid; may be useful as a marker of mitochondrial dysfunction
Catalog No. 50-289-3509