Learn More
Cayman Chemical ArachIdonyl-L-carnItInchl 25mg
SDP
Supplier:  Cayman Chemical 900093925 MG
Encompass_Preferred
An acylcarnitine formed from carnitine conjugated to arachidonic acid; may be useful as a marker of mitochondrial dysfunction
Catalog No. 50-289-3510