Learn More
Cayman Chemical ArachIdonyl m-NItronIlIn 10mg
SDP
Supplier:  Cayman Chemical 9005910 MG
Encompass_Preferred
A nitroaniline fatty acid amide used to measure FAAH activity; exposure to FAAH activity results in the release of the yellow colorimetric dye m-nitroaniline (ε = 13,500 at 410 nm) allowing the fast and convenient measurement of FAAH activity using a 96-well plate spectrophotometer
Catalog No. 50-289-5649