Learn More
Cayman Chemical Aralosde A 5mg
SDP
Supplier:  Cayman Chemical 302195 MG
Encompass_Preferred
A triterpenoid saponin with anti-inflammatory and gastroprotective activities; increases apoptosis of and decreases production of IL-1β and IL-6 in human MH7A rheumatoid arthritis fibroblast-like synoviocytes from 4-16 µM; reduces gastric acid secretion and lesion size in rat models of hydrochloric acid and ethanol-, aspirin-, water immersion stress-, or pyloric ligation-induced ulcer formation at 50 and 100 mg/kg
Catalog No. 50-298-8306