Learn More
Medchemexpress LLC Araloside VII (Congmunoside VII) | 340982-22-1 | MFCD22124709 | 99.9% | 1121.26 g/mol | C54H88O24 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN200210MG
Encompass_Preferred
Araloside VII is a naturally occurring saponin isolated from leaves of Aralias elate, supplied for research use. It is provided as a solid, white to yellow powder with a molecular formula of C54H88O24, molecular weight ≈1121.26 g/mol, and CAS number 340982-22-1. Store protected from light at 4°C; in solution, store at -80°C for long-term (up to 6 months) or -20°C for short-term (about 1 month).
  • natural saponin isolated from Aralias elate leaves.
  • high purity (≈99.9%) for research applications.
  • molecular weight ≈1121.26 g/mol; formula C54H88O24.
  • provided as a 10 mg solid suitable for small-scale experiments.
  • store protected from light at 4°C; use cold solvent storage for extended stability.
  • intended for laboratory research use only; not for human or diagnostic use.
Catalog No. 50-002-95783