Learn More
Medchemexpress LLC Icomidocholic acid | 246529-22-6 | 99.98% | 702.10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY197965MG
Encompass_Preferred
Icomidocholic acid (Aramchol) is a lipid molecule synthesized from cholic acid and arachidic acid. It is an orally active SCD1 inhibitor and cholesterol solubilizer with antifibrotic effects. It is used in the study of nonalcoholic fatty liver disease (NAFLD) and nonalcoholic steatohepatitis (NASH).
  • Reduces liver fat content
  • Dissolves cholesterol crystals
  • Prevents gallstone formation
  • For research use only
Catalog No. 50-197-0036