Learn More
Medchemexpress LLC Arbutin (β-Arbutin) | 497-76-7 | 99.8% | 272.25 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYN0192
Encompass_Preferred
Arbutin (β-Arbutin) is a competitive inhibitor of tyrosinase and is used as a depigmenting agent. It is a natural polyphenol isolated from the bearberry plant Arctostaphylos uvaursi, possessing anti-oxidant, anti-inflammatory, and anti-tumor properties.
  • Inhibits the viability of B16 murine melanoma cells in a time- and dose-dependent manner.
  • Increases the apoptosis rate of B16 murine melanoma cells.
  • Exhibits protective effects on isoproterenol-induced cardiac hypertrophy in mice.
  • Ameliorates isoproterenol-induced myocardial injury.
Catalog No. 50-004-29516