Learn More
Selleck Chemical LLC Arbutin 25mg 497-76-7 Uva,p-Arbutin, -Arbutin
SDP
Supplier:  Selleck Chemical LLC S226325MG
Encompass_Preferred
Arbutin (Uva, p-Arbutin, ?-Arbutin) is a tyrosinase inhibitor with IC50 of 0.9 mM and 0.7 mM for Monophenolase and Diphenolase, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-582-2