Learn More
Fauske & Associates, a Division of Stone & Webster, LLC ARC TEST CELL
Supplier:  Fauske & Associates, a Division of Stone & Webster, LLC ARCFS2BC
Encompass
"ARC Test Cell -- all metal; long neck, 316SS;1"" I.D.; .035 wall with 1/4"" O.D. x .035 wall x 1"" stems; bottom clip;volume: 9.5 mL; can be used with the Accelerating Rate Calorimeter(ARC) and the ARSST"
Catalog No. NC1951864