Learn More
Medchemexpress LLC Arc186 sodium | 1 MG

Supplier: Medchemexpress LLC HY153098A1MG
ARC186 sodium is an unconjugated 40KDa PEG aptamer with a sequence identical to ARC1905 sodium. This highly potent complement inhibitor functions by blocking the convertase-catalyzed activation of C5.
- Purity: 94.13%
- Appearance: Solid
- Color: White to off-white
- Active in immunology and inflammation pathways
- Inhibits the complement system
- Relevant for inflammation or immune system disease research
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.