Learn More
Cayman Chemical ArcaInsulfate 100mg
SDP
Supplier:  Cayman Chemical 33284100 MG
Encompass_Preferred
An NMDA receptor antagonist (IC50 = 9.13 µM); inhibits NMDA and glycine-induced inward currents in X. laevis oocytes expressing rat NMDA receptors (IC50 = 60.1 µM); impairs contextual and auditory fear conditioning in rats when administered post-training at 10 mg/kg, i.p.
Catalog No. 50-295-0225